Gene Transcripts

Gene Symbol
ANO8
Entrez Gene ID
57719
Gene Name
anoctamin 8
Gene Aliases
KIAA1623, TMEM16H
Chromosome Location
Chr. 19: 17323223 - 17334834 on Build GRCh38
UniGene ID
Hs.590990
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020959.2 NP_066010.1 7 881
XM_017027048.1 XP_016882537.1 7 886
GenBank mRNA AB046843.2 - 7 881

View Full Product Details