Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127173.1 | NP_001120645.1 | 4 | 549 | |
NM_021189.3 | NP_067012.1 | 5 | 688 | ||
GenBank mRNA | AF062733.2 | - | 5 | 645 | |
AF363367.1 | - | 4 | 547 | ||
AF529206.1 | - | 4 | 414 | ||
AK289541.1 | - | 5 | 688 | ||
AK308915.1 | - | 5 | 660 | ||
AY046418.1 | - | 4 | 452 | ||
AY358332.1 | - | 4 | 553 | ||
BC033819.1 | - | 4 | 497 | 5199627 |