Gene Transcripts

Gene Symbol
NTN4
Entrez Gene ID
59277
Gene Name
netrin 4
Gene Aliases
PRO3091
Chromosome Location
Chr. 12: 95657807 - 95790758 on Build GRCh38
UniGene ID
Hs.201034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021229.3 NP_067052.2 3 1170
GenBank mRNA AF119916.3 - 4 707
AF278532.1 - 3 1172
AF297711.1 - 3 1022
AK024691.1 - 2 246
AK226070.1 - 3 1027
AK290591.1 - 3 707
AK315796.1 - 3 1029
AL137540.1 - 1 38
AL833767.1 - 2 574
AY330211.1 - 3 721
BC013591.1 - 2 245
BC136798.1 - 3 1145 9020788
BC136799.1 - 3 1145 9020789

View Full Product Details