Gene Transcripts

Gene Symbol
FASTKD5
Entrez Gene ID
60493
Gene Name
FAST kinase domains 5
Gene Aliases
dJ1187M17.5
Chromosome Location
Chr. 20: 3146519 - 3159886 on Build GRCh38
UniGene ID
Hs.741168
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021826.4 NP_068598.1 2 927
GenBank mRNA AB058695.1 - 1 3293
AK023211.1 - 2 906
AK301748.1 - 2 879
BC007413.2 - 2 927 3010170
BC011856.1 - 2 891 4298806

View Full Product Details