Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024504.3 | NP_078780.1 | 6 | 1405 | |
XM_011517572.1 | XP_011515874.1 | 5 | 685 | ||
GenBank mRNA | AF319458.1 | - | 6 | 1405 | |
AK022595.1 | - | 6 | 1405 | ||
BC052311.1 | - | 6 | 1318 | 6671984 | |
KJ463837.1 | - | 5 | 1203 |