Gene Transcripts

Gene Symbol
ERAP2
Entrez Gene ID
64167
Gene Name
endoplasmic reticulum aminopeptidase 2
Gene Aliases
L-RAP, LRAP
Chromosome Location
Chr. 5: 96875940 - 96919703 on Build GRCh38
UniGene ID
Hs.482910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011543544.1 XP_011541846.1 15 2553
GenBank mRNA AB109031.1 - 16 2589
AB163917.1 - 16 2690
AF191545.1 - 15 2561
AK312864.1 - 16 2602
AY028805.1 - 15 2575
BC065240.1 - 15 2448 6166289

View Full Product Details