Gene Transcripts
- Gene Symbol
- FNDC3B
- Entrez Gene ID
- 64778
- Gene Name
- fibronectin type III domain containing 3B
- Gene Aliases
- FAD104, PRO4979, YVTM2421
- Chromosome Location
- Chr. 3: 172039628 - 172400703 on Build GRCh38
- UniGene ID
- Hs.744888
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135095.1 |
NP_001128567.1
|
18, 19 |
2183 |
|
NM_022763.3 |
NP_073600.3
|
18, 19 |
2256 |
|
XM_017007062.1 |
XP_016862551.1
|
17, 18 |
1954 |
|
XM_017007063.1 |
XP_016862552.1
|
13, 14 |
1440 |
|
GenBank mRNA |
AB098597.1 |
- |
17, 18 |
2141 |
|
AK027052.1 |
- |
2, 3 |
239 |
|
AK075220.1 |
- |
8, 9 |
878 |
|
AK127826.1 |
- |
18, 19 |
2236 |
|
AK223599.1 |
- |
18, 19 |
2287 |
|
AK314478.1 |
- |
18, 19 |
2183 |
|
AY358146.1 |
- |
11, 12 |
1129 |
|
BC033635.2 |
- |
6, 7 |
731 |
|
BC039297.1 |
- |
18, 19 |
2147 |
5297602 |
BX648340.1 |
- |
18, 19 |
2178 |
|
View Full Product Details