Gene Transcripts

Gene Symbol
GINS3
Entrez Gene ID
64785
Gene Name
GINS complex subunit 3
Gene Aliases
PSF3
Chromosome Location
Chr. 16: 58392394 - 58406144 on Build GRCh38
UniGene ID
Hs.47125
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126129.1 NP_001119601.1 4 909
NM_001126130.1 NP_001119602.1 2 558
NM_022770.3 NP_073607.2 3 792
GenBank mRNA AK023974.1 - 2 464
AK127454.1 - 1 1721
AK315622.1 - 3 743
AL137379.1 - 1 128
BC005879.1 - 3 667 3529058
BC014437.1 - 3 686 4870865
CR457283.1 - 2 350
KU178752.1 - 3 584
KU178753.1 - 4 701
KU178754.1 - 4 643
KU178755.1 - 2 350

View Full Product Details