Gene Transcripts

Gene Symbol
PAPOLG
Entrez Gene ID
64895
Gene Name
poly(A) polymerase gamma
Gene Aliases
Chromosome Location
Chr. 2: 60756230 - 60802086 on Build GRCh38
UniGene ID
Hs.387471
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022894.3 NP_075045.2 7, 8 851
XM_005264500.3 XP_005264557.1 7, 8 851
XM_005264501.1 XP_005264558.1 7, 8 669
GenBank mRNA AF312211.1 - 7, 8 833
AJ308101.1 - 7, 8 602
AK021867.1 - 7, 8 787
AK023544.1 - 6, 7 600
AK314663.1 - 7, 8 815
AY029162.1 - 7, 8 611
BC016738.1 - 7, 8 822
BC110906.1 - 7, 8 790
BC111701.1 - 7, 8 725 40038554
BC113747.1 - 7, 8 725 40038563

View Full Product Details