Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_203365.3 | NP_976241.1 | 4 | 503 | |
NM_213589.2 | NP_998754.1 | 4 | 503 | ||
XM_005246777.3 | XP_005246834.1 | 4 | 498 | ||
XM_005246779.4 | XP_005246836.1 | 4 | 1397 | ||
XM_006712695.3 | XP_006712758.1 | 5 | 1478 | ||
XM_011511646.2 | XP_011509948.1 | 5 | 570 | ||
XM_011511647.1 | XP_011509949.1 | 3 | 278 | ||
XM_011511648.1 | XP_011509950.1 | 3 | 278 | ||
XM_011511649.2 | XP_011509951.1 | 4 | 498 | ||
GenBank mRNA | AB051468.1 | - | 3 | 236 | |
AB053311.1 | - | 4 | 497 | ||
AJ584699.1 | - | 4 | 497 | ||
AK096270.1 | - | 4 | 447 | ||
AY494951.1 | - | 4 | 499 | ||
AY523977.1 | - | 4 | 499 | ||
AY523978.1 | - | 4 | 499 |