Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000394.3 | NP_000385.1 | 2 | 303 | |
XM_005261093.3 | XP_005261150.1 | 2 | 408 | ||
GenBank mRNA | BC113598.1 | - | 2 | 232 | |
BX118596.1 | - | 2 | 303 | ||
CR407691.1 | - | 2 | 211 | ||
KM220588.1 | - | 2 | 261 | ||
KM220590.1 | - | 2 | 268 | ||
KM220591.1 | - | 2 | 227 | ||
KM220592.1 | - | 2 | 269 | ||
U05569.1 | - | 2 | 278 | ||
U66584.1 | - | 2 | 211 | ||
X14789.1 | - | 1 | 1452 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001314050.2 | NP_001300979.1 | 2 | 270 | |
NM_001320719.1 | NP_001307648.1 | 2 | 196 | ||
GenBank mRNA | BC069528.1 | - | 2 | 261 | |
BQ640485.1 | - | 2 | 196 | ||
KM220589.1 | - | 2 | 266 |