Gene Transcripts

Gene Symbol
CUEDC2
Entrez Gene ID
79004
Gene Name
CUE domain containing 2
Gene Aliases
C10orf66, bA18I14.5
Chromosome Location
Chr. 10: 102423245 - 102432666 on Build GRCh38
UniGene ID
Hs.500874
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024040.2 NP_076945.2 5 509
GenBank mRNA AB025425.1 - 4 358
AB232358.1 - 4 358
BC000262.1 - 5 439 3353174
CX165866.1 - 5 393

View Full Product Details