Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080432.2 | NP_001073901.1 | 9 | 1618 | |
XM_011523313.2 | XP_011521615.1 | 9 | 1612 | ||
GenBank mRNA | AB051539.1 | - | 9 | 1402 | |
AK096554.1 | - | 9 | 1425 | ||
AK295196.1 | - | 8 | 771 | ||
BC003583.1 | - | 3 | 129 | ||
BC030798.1 | - | 3 | 386 | ||
BC132892.1 | - | 4 | 321 | ||
BC137091.1 | - | 4 | 301 |