Gene Transcripts

Gene Symbol
FSD1
Entrez Gene ID
79187
Gene Name
fibronectin type III and SPRY domain containing 1
Gene Aliases
GLFND, MIR1
Chromosome Location
Chr. 19: 4304594 - 4323846 on Build GRCh38
UniGene ID
Hs.28144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024333.2 NP_077309.1 2 231
XM_005259644.2 XP_005259701.1 2 230
GenBank mRNA AF316829.1 - 2 122
AK021750.1 - 2 131
AK295523.1 - 2 126
AK315661.1 - 2 225
AY032617.1 - 2 154
BC003124.1 - 2 131 3502614

View Full Product Details