Gene Transcripts

Gene Symbol
CRYGD
Entrez Gene ID
1421
Gene Name
crystallin gamma D
Gene Aliases
CACA, CCA3, CCP, CRYG4, CTRCT4, PCC, cry-g-D
Chromosome Location
Chr. 2: 208121607 - 208124589 on Build GRCh38
UniGene ID
Hs.546247
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006891.3 NP_008822.2 3 467
GenBank mRNA BC117338.1 - 3 467
BC117340.1 - 3 467 40125891
CD674533.1 - 2 331
EL946552.1 - 3 383
KT966392.1 - 3 375
U66583.1 - 2 351

View Full Product Details