Gene Transcripts

Gene Symbol
CSNK1G2
Entrez Gene ID
1455
Gene Name
casein kinase 1 gamma 2
Gene Aliases
CK1g2
Chromosome Location
Chr. 19: 1941149 - 1981338 on Build GRCh38
UniGene ID
Hs.651905
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319.6 NP_001310.3 9 1418
XM_005259498.1 XP_005259555.1 8 1520
XM_005259499.3 XP_005259556.1 8 1327
XM_005259500.1 XP_005259557.1 9 1430
XM_005259501.2 XP_005259558.1 9 1804
XM_017026297.1 XP_016881786.1 8 1394
XM_017026298.1 XP_016881787.1 9 1804
GenBank mRNA AB451278.1 - 8 895
AB451410.1 - 8 895
AF001177.1 - 9 2140
AY007134.1 - 4 315
BC018693.2 - 9 1391 3010717
BC018699.2 - 9 1391 3010717
BC020972.1 - 8 948 3851613
BT009922.1 - 8 895
U89896.1 - 8 1134

View Full Product Details