Gene Transcripts

Gene Symbol
NOL10
Entrez Gene ID
79954
Gene Name
nucleolar protein 10
Gene Aliases
PQBP5
Chromosome Location
Chr. 2: 10570766 - 10689991 on Build GRCh38
UniGene ID
Hs.222494
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001261392.1 NP_001248321.1 5 426
NM_001261394.1 NP_001248323.1 6 504
NM_024894.3 NP_079170.2 6 504
NR_048552.1 5 458
XM_011510400.2 XP_011508702.1 6 492
XM_011510402.2 XP_011508704.1 6 492
XM_017004990.1 XP_016860479.1 4 246
GenBank mRNA AK024137.1 - 6 489
AK290680.1 - 6 468
AK297169.1 - 5 414
AK299617.1 - 5 446
BC005125.2 - 6 484 3841643
BC037841.1 - 1 1564
DC346574.1 - 6 504

View Full Product Details