Gene Transcripts

Gene Symbol
NOL10
Entrez Gene ID
79954
Gene Name
nucleolar protein 10
Gene Aliases
PQBP5
Chromosome Location
Chr. 2: 10570766 - 10689991 on Build GRCh38
UniGene ID
Hs.222494
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001261394.1 NP_001248323.1 4 360
NM_024894.3 NP_079170.2 4 360
NR_048552.1 3 314
XM_011510398.2 XP_011508700.1 4 364
XM_011510399.2 XP_011508701.1 3 302
XM_011510400.2 XP_011508702.1 4 348
XM_011510402.2 XP_011508704.1 4 348
XM_017004990.1 XP_016860479.1 2 102
GenBank mRNA AK024000.1 - 4 338
AK024137.1 - 4 345
AK290680.1 - 4 324
AK299617.1 - 3 302
BC005125.2 - 4 340 3841643
DC346574.1 - 4 360

View Full Product Details