Gene Transcripts
- Gene Symbol
- CTC1
- Entrez Gene ID
- 80169
- Gene Name
- CTS telomere maintenance complex component 1
- Gene Aliases
- AAF-132, AAF132, C17orf68, CRMCC, tmp494178
- Chromosome Location
- Chr. 17: 8224821 - 8248095 on Build GRCh38
- UniGene ID
- Hs.156055
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025099.5 |
NP_079375.3
|
14, 15 |
2544 |
|
NR_046431.1 |
|
14, 15 |
2439 |
|
XM_006721577.3 |
XP_006721640.1
|
13, 14 |
2399 |
|
XM_006721578.3 |
XP_006721641.1
|
14, 15 |
2528 |
|
XM_011524010.2 |
XP_011522312.1
|
14, 15 |
2423 |
|
XM_011524011.2 |
XP_011522313.1
|
13, 14 |
2293 |
|
GenBank mRNA |
AK025823.1 |
- |
4, 5 |
687 |
|
AK091077.1 |
- |
14, 15 |
2495 |
|
AK225375.1 |
- |
4, 5 |
687 |
|
AK310636.1 |
- |
3, 4 |
713 |
|
AL831955.1 |
- |
14, 15 |
2386 |
|
BC026057.2 |
- |
5, 6 |
685 |
|
BC110373.1 |
- |
6, 7 |
911 |
|
BC111783.1 |
- |
14, 15 |
2495 |
40054070 |
DQ451688.1 |
- |
14, 15 |
2485 |
|
FJ790670.1 |
- |
14, 15 |
2485 |
|
View Full Product Details