Gene Transcripts
- Gene Symbol
- TRAF3IP3
- Entrez Gene ID
- 80342
- Gene Name
- TRAF3 interacting protein 3
- Gene Aliases
- T3JAM
- Chromosome Location
- Chr. 1: 209756032 - 209782323 on Build GRCh38
- UniGene ID
- Hs.147434
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001287754.1 |
NP_001274683.1
|
5, 6 |
658 |
|
NM_001320143.1 |
NP_001307072.1
|
12, 13 |
1703 |
|
NM_001320144.1 |
NP_001307073.1
|
12, 13 |
1564 |
|
NM_025228.3 |
NP_079504.2
|
12, 13 |
1624 |
|
XM_005273279.4 |
XP_005273336.1
|
5, 6 |
2337 |
|
XM_005273280.2 |
XP_005273337.1
|
5, 6 |
600 |
|
XM_011510018.2 |
XP_011508320.1
|
12, 13 |
1608 |
|
XM_017002399.1 |
XP_016857888.1
|
12, 13 |
1608 |
|
GenBank mRNA |
AK122792.1 |
- |
12, 13 |
1607 |
|
AK125601.1 |
- |
12, 13 |
1703 |
|
AK225762.1 |
- |
12, 13 |
1519 |
|
AK312755.1 |
- |
12, 13 |
1518 |
|
AL049667.1 |
- |
12, 13 |
1417 |
|
BC130417.1 |
- |
11, 12 |
1304 |
40146448 |
BC144139.1 |
- |
11, 12 |
1304 |
9052659 |
BX648450.1 |
- |
12, 13 |
1603 |
|
View Full Product Details