Gene Transcripts

Gene Symbol
CDT1
Entrez Gene ID
81620
Gene Name
chromatin licensing and DNA replication factor 1
Gene Aliases
DUP, RIS2
Chromosome Location
Chr. 16: 88803778 - 88809258 on Build GRCh38
UniGene ID
Hs.122908
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030928.3 NP_112190.2 7 1061
GenBank mRNA AB053172.1 - 7 1061
AF070552.1 - 5 600
AF321125.1 - 7 1032
BC000137.2 - 7 1023 3048353
BC008676.1 - 7 899
BC008860.2 - 7 954
BC009410.1 - 7 1017 3613566
BC014202.2 - 3 322
BC021126.1 - 5 593
BC049205.1 - 5 665

View Full Product Details