Gene Transcripts

Gene Symbol
TCF7L1
Entrez Gene ID
83439
Gene Name
transcription factor 7 like 1
Gene Aliases
TCF-3, TCF3
Chromosome Location
Chr. 2: 85133460 - 85310388 on Build GRCh38
UniGene ID
Hs.516297
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031283.2 NP_112573.1 3 560
XM_006712109.2 XP_006712172.1 3 350
GenBank mRNA AB031046.1 - 3 409
BC058894.1 - 3 569 6141641

View Full Product Details