Gene Transcripts

Gene Symbol
EMILIN2
Entrez Gene ID
84034
Gene Name
elastin microfibril interfacer 2
Gene Aliases
EMILIN-2, FOAP-10
Chromosome Location
Chr. 18: 2847030 - 2914092 on Build GRCh38
UniGene ID
Hs.532815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032048.2 NP_114437.2 4 1100
XM_017026038.1 XP_016881527.1 4 1087
GenBank mRNA AF270513.1 - 4 1100
AL552384.3 - 4 870
BC136541.1 - 4 1054 9020531

View Full Product Details