Gene Transcripts

Gene Symbol
ADGRV1
Entrez Gene ID
84059
Gene Name
adhesion G protein-coupled receptor V1
Gene Aliases
FEB4, GPR98, MASS1, USH2B, USH2C, VLGR1, VLGR1b
Chromosome Location
Chr. 5: 90558681 - 91164226 on Build GRCh38
UniGene ID
Hs.591777
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032119.3 NP_115495.3 14 2725
NR_003149.1 14 2725
XM_017009963.1 XP_016865452.1 14 2844
XM_017009964.1 XP_016865453.1 14 2844
XM_017009965.1 XP_016865454.1 14 2726
XM_017009966.1 XP_016865455.1 14 2844
XM_017009967.1 XP_016865456.1 13 2633
XM_017009968.1 XP_016865457.1 14 2844
XM_017009969.1 XP_016865458.1 14 2844
XM_017009970.1 XP_016865459.1 14 2844
XM_017009971.1 XP_016865460.1 14 2844
XM_017009974.1 XP_016865463.1 14 2844
GenBank mRNA AB075823.1 - 7 1395
AF435925.1 - 14 2725

View Full Product Details