Gene Transcripts

Gene Symbol
ASCC2
Entrez Gene ID
84164
Gene Name
activating signal cointegrator 1 complex subunit 2
Gene Aliases
ASC1p100, p100
Chromosome Location
Chr. 22: 29788608 - 29838304 on Build GRCh38
UniGene ID
Hs.731754
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242906.1 NP_001229835.1 15 1636
NM_032204.4 NP_115580.2 16 1864
XM_005261775.2 XP_005261832.1 17 1904
XM_011530442.2 XP_011528744.1 17 1904
XM_011530443.2 XP_011528745.1 18 1970
XM_011530444.2 XP_011528746.1 18 2017
XM_011530445.2 XP_011528747.1 17 1951
XM_011530446.2 XP_011528748.1 16 1834
XM_011530448.2 XP_011528750.1 16 1737
XM_011530449.2 XP_011528751.1 15 1675
XM_011530450.2 XP_011528752.1 18 2020
XM_011530451.2 XP_011528753.1 19 2094
XM_011530452.2 XP_011528754.1 18 2067
XM_011530453.2 XP_011528755.1 17 1922
XM_011530454.2 XP_011528756.1 16 1806
XM_011530455.2 XP_011528757.1 15 1735
XM_017028991.1 XP_016884480.1 16 4042
XM_017028992.1 XP_016884481.1 16 4042
XM_017028993.1 XP_016884482.1 15 3967
XM_017028994.1 XP_016884483.1 15 3967
XM_017028995.1 XP_016884484.1 15 3883
XM_017028996.1 XP_016884485.1 17 2844
XM_017028997.1 XP_016884486.1 15 3883
XM_017028998.1 XP_016884487.1 17 2844
XM_017028999.1 XP_016884488.1 18 1970
XM_017029000.1 XP_016884489.1 17 1951
XM_017029001.1 XP_016884490.1 16 1737
XM_017029002.1 XP_016884491.1 15 1675
XM_017029003.1 XP_016884492.1 17 1811
XM_017029004.1 XP_016884493.1 14 1600
XM_017029005.1 XP_016884494.1 18 2020
XM_017029006.1 XP_016884495.1 19 2094
XM_017029007.1 XP_016884496.1 17 1922
XM_017029008.1 XP_016884497.1 16 1806
XM_017029009.1 XP_016884498.1 15 1735
GenBank mRNA AK022886.1 - 18 1861
AK025241.1 - 8 893
AK222723.1 - 8 893
AK223033.1 - 8 892
AK303257.1 - 15 1606
BC025368.2 - 16 1825 3955747
BU508026.1 - 3 214

View Full Product Details