Gene Transcripts

Gene Symbol
SLC37A3
Entrez Gene ID
84255
Gene Name
solute carrier family 37 member 3
Gene Aliases
Chromosome Location
Chr. 7: 140333752 - 140404352 on Build GRCh38
UniGene ID
Hs.446021
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287498.1 NP_001274427.1 10 1115
NM_032295.3 NP_115671.1 10 1115
NM_207113.2 NP_996996.1 10 1115
XM_011516626.2 XP_011514928.1 12 1612
XM_011516627.2 XP_011514929.1 12 1611
XM_017012712.1 XP_016868201.1 12 1588
XM_017012713.1 XP_016868202.1 12 1607
XM_017012714.1 XP_016868203.1 10 1385
XM_017012715.1 XP_016868204.1 10 1367
XM_017012716.1 XP_016868205.1 10 1371
GenBank mRNA AK074823.1 - 10 1076
AK299276.1 - 11 1161
AK300265.1 - 9 1445
AL136583.1 - 10 1022
BC028380.1 - 10 1065 4811814
BC046567.1 - 10 1067 5246993

View Full Product Details