Gene Transcripts

Gene Symbol
RAB11FIP4
Entrez Gene ID
84440
Gene Name
RAB11 family interacting protein 4
Gene Aliases
RAB11-FIP4
Chromosome Location
Chr. 17: 31391624 - 31538217 on Build GRCh38
UniGene ID
Hs.406788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303542.2 NP_001290471.2 9 1269
NM_032932.5 NP_116321.2 11 1562
GenBank mRNA AB058724.1 - 11 1251
AJ314646.2 - 11 1251
AK074341.1 - 4 340
AK097424.1 - 9 1212
AL831830.1 - 3 311
AY169244.1 - 11 1562
BC093914.1 - 11 1365 7939759
BC101517.1 - 11 1365 8069023

View Full Product Details