Gene Transcripts

Gene Symbol
PLA2G12B
Entrez Gene ID
84647
Gene Name
phospholipase A2 group XIIB
Gene Aliases
FKSG71, GXIIB, GXIIIsPLA2, PLA2G13, sPLA2-GXIIB
Chromosome Location
Chr. 10: 72934762 - 72955044 on Build GRCh38
UniGene ID
Hs.333175
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318124.1 NP_001305053.1 3 487
NM_001318125.1 NP_001305054.1 2 398
NM_032562.4 NP_115951.2 3 487
XM_005270244.4 XP_005270301.1 3 700
GenBank mRNA AF339053.1 - 3 446
AF349540.1 - 3 390
AK311349.1 - 2 385
AK313712.1 - 3 450
AY358032.1 - 3 420
BC093996.1 - 3 409 7939841
BC111946.1 - 3 409 8327414
BC143532.1 - 3 409 9052041
BP274516.1 - 3 487
DA536331.1 - 2 385

View Full Product Details