Gene Transcripts

Gene Symbol
MS4A14
Entrez Gene ID
84689
Gene Name
membrane spanning 4-domains A14
Gene Aliases
MS4A16, NYD-SP21
Chromosome Location
Chr. 11: 60396014 - 60417756 on Build GRCh38
UniGene ID
Hs.709736
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079692.2 NP_001073160.1 4 2157
NM_001261827.1 NP_001248756.1 5 2256
NM_001261828.1 NP_001248757.1 6 2307
NM_032597.4 NP_115986.3 5 2208
NR_049731.1 6 2377
NR_049732.1 6 2329
NR_049733.1 5 2278
NR_049734.1 7 2428
GenBank mRNA AF367473.2 - 5 1719
AK057418.1 - 6 1923
AK314674.1 - 5 1918
AL137391.1 - 1 1075
AY094610.1 - 5 1792
AY094611.1 - 4 1741
AY584610.1 - 4 1741

View Full Product Details