Gene Transcripts

Gene Symbol
ATAD1
Entrez Gene ID
84896
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
AFDC1, FNP001, THORASE
Chromosome Location
Chr. 10: 87751512 - 87841345 on Build GRCh38
UniGene ID
Hs.435948
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321967.1 NP_001308896.1 4 355
NM_001321968.1 NP_001308897.1 4 355
NM_001321969.1 NP_001308898.1 4 355
NM_032810.3 NP_116199.2 4 727
NR_135914.1 4 355
XM_005270252.4 XP_005270309.1 4 449
XM_011540302.1 XP_011538604.1 2 120
XM_011540303.2 XP_011538605.1 4 402
XM_017016847.1 XP_016872336.1 4 596
XM_017016848.1 XP_016872337.1 4 689
XM_017016849.1 XP_016872338.1 4 478
XM_017016850.1 XP_016872339.1 4 529
XM_017016851.1 XP_016872340.1 4 402
GenBank mRNA AF361493.1 - 3 561
AK075223.1 - 4 656
AK304297.1 - 2 118
AL834370.1 - 3 140
BC063530.1 - 3 186
BC073998.1 - 3 298 6199813

View Full Product Details