Gene Transcripts

Gene Symbol
ATAD1
Entrez Gene ID
84896
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
AFDC1, FNP001, THORASE
Chromosome Location
Chr. 10: 87751512 - 87841345 on Build GRCh38
UniGene ID
Hs.435948
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321967.1 NP_001308896.1 6 745
NM_001321968.1 NP_001308897.1 6 745
NM_001321969.1 NP_001308898.1 7 835
NM_032810.3 NP_116199.2 6 1117
NR_135914.1 6 745
XM_005270252.4 XP_005270309.1 6 839
XM_011540302.1 XP_011538604.1 4 510
XM_011540303.2 XP_011538605.1 6 792
XM_017016847.1 XP_016872336.1 6 986
XM_017016848.1 XP_016872337.1 6 1079
XM_017016849.1 XP_016872338.1 7 958
XM_017016850.1 XP_016872339.1 7 1009
GenBank mRNA AF361493.1 - 5 951
AK027506.1 - 3 345
AK075223.1 - 6 1046
AK304297.1 - 4 508
AL834370.1 - 5 530
BC010868.1 - 2 180
BC063530.1 - 5 576
BC073998.1 - 5 688 6199813

View Full Product Details