Gene Transcripts

Gene Symbol
PLXDC2
Entrez Gene ID
84898
Gene Name
plexin domain containing 2
Gene Aliases
TEM7R
Chromosome Location
Chr. 10: 19816215 - 20289856 on Build GRCh38
UniGene ID
Hs.658134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032812.8 NP_116201.7 3, 4 1118
XM_011519750.2 XP_011518052.1 3, 4 1346
GenBank mRNA AF378757.1 - 3, 4 490
AK304591.1 - 4, 5 1002
AY358486.1 - 3, 4 1118

View Full Product Details