Gene Transcripts

Gene Symbol
DCX
Entrez Gene ID
1641
Gene Name
doublecortin
Gene Aliases
DBCN, DC, LISX, SCLH, XLIS
Chromosome Location
Chr. X: 111293779 - 111412232 on Build GRCh38
UniGene ID
Hs.34780
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000555.3 NP_000546.2 3, 4 1125
NM_001195553.1 NP_001182482.1 3, 4 827
NM_178151.2 NP_835364.1 3, 4 954
NM_178152.2 NP_835365.1 3, 4 827
NM_178153.2 NP_835366.1 3, 4 827
XM_011530878.2 XP_011529180.1 3, 4 799
XM_011530879.2 XP_011529181.1 3, 4 799
XM_011530880.2 XP_011529182.1 3, 4 799
XM_017029312.1 XP_016884801.1 3, 4 892
GenBank mRNA AF034634.1 - 2, 3 710
AF040254.1 - 3, 4 752
AF040255.1 - 3, 4 752
AJ003112.1 - 3, 4 1125
AK290455.1 - 3, 4 774
AK297297.1 - 3, 4 928
BC027925.1 - 3, 4 769 5240179

View Full Product Details