Gene Transcripts

Gene Symbol
MBD3L1
Entrez Gene ID
85509
Gene Name
methyl-CpG binding domain protein 3 like 1
Gene Aliases
MBD3L
Chromosome Location
Chr. 19: 8842593 - 8843340 on Build GRCh38
UniGene ID
Hs.133912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145208.2 NP_660209.2 1 615
GenBank mRNA AB451462.1 - 1 529
AY038022.1 - 1 552
BC112058.1 - 1 615 8327526
BC112064.1 - 1 615 8327532

View Full Product Details