Gene Transcripts
- Gene Symbol
- GTPBP10
- Entrez Gene ID
- 85865
- Gene Name
- GTP binding protein 10
- Gene Aliases
- ObgH2, UG0751c10
- Chromosome Location
- Chr. 7: 90346665 - 90391455 on Build GRCh38
- UniGene ID
- Hs.593547
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042717.2 |
NP_001036182.1
|
3, 4 |
374 |
|
NM_033107.3 |
NP_149098.2
|
5, 6 |
611 |
|
GenBank mRNA |
AF351613.1 |
- |
3, 4 |
375 |
|
AK095561.1 |
- |
5, 6 |
601 |
|
AK294325.1 |
- |
3, 4 |
324 |
|
AL525056.3 |
- |
5, 6 |
553 |
|
BC004923.2 |
- |
4, 5 |
508 |
|
BC021573.1 |
- |
4, 5 |
508 |
|
BC107714.1 |
- |
5, 6 |
595 |
6714887 |
BC144162.1 |
- |
3, 4 |
321 |
|
CR933597.1 |
- |
4, 5 |
425 |
|
View Full Product Details