Gene Transcripts

Gene Symbol
NAV1
Entrez Gene ID
89796
Gene Name
neuron navigator 1
Gene Aliases
POMFIL3, STEERIN1, UNC53H1
Chromosome Location
Chr. 1: 201648322 - 201826974 on Build GRCh38
UniGene ID
Hs.585374
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167738.1 NP_001161210.1 15 2715
NM_020443.4 NP_065176.3 18 4055
XM_006711609.2 XP_006711672.2 16 2690
XM_006711610.2 XP_006711673.2 15 2666
XM_006711611.1 XP_006711674.1 16 2806
XM_011510097.1 XP_011508399.1 17 3725
XM_011510098.1 XP_011508400.1 17 3710
XM_011510099.1 XP_011508401.1 16 3701
XM_011510100.1 XP_011508402.1 17 3563
XM_011510101.1 XP_011508403.1 16 3539
XM_011510102.1 XP_011508404.1 15 3530
XM_017002751.1 XP_016858240.1 16 3554
XM_017002752.1 XP_016858241.1 14 2657
GenBank mRNA AB032977.3 - 18 3388
AJ488101.1 - 18 3586
AK024265.1 - 3 220
AY043013.1 - 18 4055
BC007523.1 - 4 337
BX648460.1 - 15 2682

View Full Product Details