Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138300.3 | NP_612157.1 | 3 | 408 | |
GenBank mRNA | AF289598.1 | - | 3 | 160 | |
AF457208.1 | - | 3 | 202 | ||
AK092389.1 | - | 3 | 357 | ||
BC006132.1 | - | 3 | 374 | 3627860 | |
BC032099.2 | - | 3 | 374 | 4126360 |