Gene Transcripts
- Gene Symbol
- PHF21B
- Entrez Gene ID
- 112885
- Gene Name
- PHD finger protein 21B
- Gene Aliases
- BHC80L, PHF4
- Chromosome Location
- Chr. 22: 44881162 - 45009928 on Build GRCh38
- UniGene ID
- Hs.254097
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135862.2 |
NP_001129334.1
|
3, 4 |
232 |
|
NM_001242450.1 |
NP_001229379.1
|
2, 3 |
272 |
|
NM_138415.4 |
NP_612424.1
|
2, 3 |
277 |
|
NR_134535.1 |
|
2, 3 |
187 |
|
XM_006724122.1 |
XP_006724185.1
|
2, 3 |
412 |
|
XM_011529855.1 |
XP_011528157.1
|
2, 3 |
412 |
|
GenBank mRNA |
AK092243.1 |
- |
3, 4 |
232 |
|
AK297317.1 |
- |
2, 3 |
265 |
|
AK299836.1 |
- |
2, 3 |
228 |
|
BC012187.1 |
- |
2, 3 |
277 |
4634631 |
CR456448.1 |
- |
2, 3 |
159 |
|
KU178842.1 |
- |
2, 3 |
126 |
|
KU178843.1 |
- |
2, 3 |
126 |
|
KU178844.1 |
- |
2, 3 |
126 |
|
View Full Product Details