Gene Transcripts

Gene Symbol
PHF21B
Entrez Gene ID
112885
Gene Name
PHD finger protein 21B
Gene Aliases
BHC80L, PHF4
Chromosome Location
Chr. 22: 44881162 - 45009928 on Build GRCh38
UniGene ID
Hs.254097
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135862.2 NP_001129334.1 11 1035
NM_001242450.1 NP_001229379.1 10 1075
NM_001284296.1 NP_001271225.1 11 1121
NM_138415.4 NP_612424.1 10 1206
NR_134535.1 9 967
XM_006724122.1 XP_006724185.1 10 1215
GenBank mRNA AK091480.1 - 11 1121
AK092243.1 - 11 1035
AK095777.1 - 4 187
AK297317.1 - 10 1068
AK299836.1 - 9 1008
BC012187.1 - 10 1206 4634631
CR456448.1 - 10 962
KU178842.1 - 10 1055
KU178843.1 - 10 929
KU178844.1 - 9 906

View Full Product Details