Gene Transcripts

Gene Symbol
SAAL1
Entrez Gene ID
113174
Gene Name
serum amyloid A like 1
Gene Aliases
SPACIA1
Chromosome Location
Chr. 11: 18080343 - 18106091 on Build GRCh38
UniGene ID
Hs.591998
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138421.2 NP_612430.2 9 993
GenBank mRNA AB489136.1 - 9 943
AK123457.1 - 9 987
BC012010.1 - 9 975 4473003
BU150462.1 - 5 564

View Full Product Details