Gene Transcripts

Gene Symbol
SFT2D1
Entrez Gene ID
113402
Gene Name
SFT2 domain containing 1
Gene Aliases
C6orf83, pRGR1
Chromosome Location
Chr. 6: 166319728 - 166342606 on Build GRCh38
UniGene ID
Hs.487143
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145169.2 NP_660152.1 3 318
NR_130112.1 4 341
NR_130113.1 4 341
NR_130114.1 4 332
GenBank mRNA AF041429.1 - 3 252
AW964442.1 - 4 263
BC018969.1 - 3 223 3959886
BC028388.1 - 4 285
BF223532.1 - 4 236
BG706927.1 - 3 307
BI597156.1 - 4 303
DB476616.1 - 4 341

View Full Product Details