Gene Transcripts

Gene Symbol
ACAP3
Entrez Gene ID
116983
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Gene Aliases
CENTB5
Chromosome Location
Chr. 1: 1292384 - 1307925 on Build GRCh38
UniGene ID
Hs.535257
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030649.2 NP_085152.2 9 780
XM_005244715.2 XP_005244772.1 9 816
XM_011540606.2 XP_011538908.1 9 829
XM_011540607.1 XP_011538909.1 9 3253
XM_011540608.2 XP_011538910.1 8 734
XM_011540609.2 XP_011538911.1 9 830
XM_017000233.1 XP_016855722.1 9 969
GenBank mRNA AB051503.1 - 8 2445
AF411981.1 - 7 580
AK093954.1 - 9 827
BC051194.1 - 9 780 4819254

View Full Product Details