Gene Transcripts
- Gene Symbol
- DCD
- Entrez Gene ID
- 117159
- Gene Name
- dermcidin
- Gene Aliases
- AIDD, DCD-1, DSEP, HCAP, PIF
- Chromosome Location
- Chr. 12: 54644591 - 54648493 on Build GRCh38
- UniGene ID
- Hs.350570
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300854.1 |
NP_001287783.1
|
3, 4 |
396 |
|
NM_053283.3 |
NP_444513.1
|
3, 4 |
396 |
|
GenBank mRNA |
AF144011.1 |
- |
3, 4 |
266 |
|
AY590150.1 |
- |
3, 4 |
260 |
|
BC062682.1 |
- |
3, 4 |
268 |
30369929 |
BC069108.1 |
- |
3, 4 |
266 |
7216925 |
BP313072.1 |
- |
3, 4 |
268 |
|
EF503687.1 |
- |
3, 4 |
396 |
|
EF503688.1 |
- |
4, 5 |
563 |
|
EF503689.1 |
- |
3, 4 |
396 |
|
View Full Product Details