Gene Transcripts

Gene Symbol
C14orf28
Entrez Gene ID
122525
Gene Name
chromosome 14 open reading frame 28
Gene Aliases
DRIP-1, DRIP1, c14_5270
Chromosome Location
Chr. 14: 44897304 - 44907257 on Build GRCh38
UniGene ID
Hs.82098
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017923.1 NP_001017923.1 2 638
GenBank mRNA AF250395.1 - 2 622
AK126985.1 - 1 394
BC030258.1 - 1 376
BC105030.1 - 2 631 8144033
BC105032.1 - 2 631 8144035

View Full Product Details