Gene Transcripts

Gene Symbol
TTC8
Entrez Gene ID
123016
Gene Name
tetratricopeptide repeat domain 8
Gene Aliases
BBS8, RP51
Chromosome Location
Chr. 14: 88824153 - 88877996 on Build GRCh38
UniGene ID
Hs.303055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288781.1 NP_001275710.1 4 462
NM_001288782.1 NP_001275711.1 4 462
NM_001288783.1 NP_001275712.1 4 462
NM_144596.3 NP_653197.2 5 492
NM_198309.3 NP_938051.1 4 462
NM_198310.3 NP_938052.1 4 462
XM_006720035.1 XP_006720098.1 3 744
XM_006720037.2 XP_006720100.1 3 744
XM_011536432.1 XP_011534734.1 3 744
XM_011536433.1 XP_011534735.1 3 744
XM_011536434.1 XP_011534736.1 3 744
XM_011536435.2 XP_011534737.1 2 148
XM_017020977.1 XP_016876466.1 3 744
XM_017020978.1 XP_016876467.1 3 744
GenBank mRNA AK093891.1 - 3 340
AK124675.1 - 4 342
AK296077.1 - 3 323
AK315772.1 - 3 347
AL833901.1 - 3 286
AY366523.1 - 3 318
AY366524.1 - 3 318
AY373972.1 - 3 318
BC026351.1 - 3 318 4799330
BC095433.1 - 3 300 30368216
BG330023.1 - 339
BX161472.1 - 3 302
BX247959.1 - 5 621
KU178849.1 - 3 266
KU178850.1 - 3 266
KU178851.1 - 3 266
KU178853.1 - 3 266
KU178854.1 - 3 266

View Full Product Details