Gene Transcripts
- Gene Symbol
- CEACAM20
- Entrez Gene ID
- 125931
- Gene Name
- carcinoembryonic antigen related cell adhesion molecule 20
- Gene Aliases
- UNQ9366
- Chromosome Location
- Chr. 19: 44503838 - 44530100 on Build GRCh38
- UniGene ID
- Hs.689632
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001102597.2 |
NP_001096067.2
|
4, 5 |
766 |
|
NM_001102598.2 |
NP_001096068.2
|
4, 5 |
766 |
|
NM_001102599.2 |
NP_001096069.2
|
4, 5 |
766 |
|
NM_001102600.2 |
NP_001096070.2
|
4, 5 |
766 |
|
XM_011526429.2 |
XP_011524731.1
|
5, 6 |
1420 |
|
XM_011526430.2 |
XP_011524732.1
|
5, 6 |
1421 |
|
XM_011526431.2 |
XP_011524733.1
|
5, 6 |
1422 |
|
GenBank mRNA |
AY358129.1 |
- |
4, 5 |
1029 |
|
EU008600.1 |
- |
4, 5 |
766 |
|
EU008601.1 |
- |
4, 5 |
766 |
|
EU008602.1 |
- |
4, 5 |
766 |
|
EU008603.1 |
- |
4, 5 |
766 |
|
View Full Product Details