Gene Transcripts

Gene Symbol
ZNF441
Entrez Gene ID
126068
Gene Name
zinc finger protein 441
Gene Aliases
Chromosome Location
Chr. 19: 11767000 - 11784078 on Build GRCh38
UniGene ID
Hs.675132
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152355.2 NP_689568.2 4 1161
GenBank mRNA AK095956.1 - 5 1134
BC144217.1 - 3 957
BX649086.1 - 1 303

View Full Product Details