Gene Transcripts

Gene Symbol
AXDND1
Entrez Gene ID
126859
Gene Name
axonemal dynein light chain domain containing 1
Gene Aliases
C1orf125
Chromosome Location
Chr. 1: 179365705 - 179554735 on Build GRCh38
UniGene ID
Hs.658505
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144696.5 NP_653297.3 17 2261
NR_073544.1 16 1900
XM_011509165.1 XP_011507467.1 17 2240
XM_011509166.2 XP_011507468.1 17 2287
XM_011509167.2 XP_011507469.1 17 2402
XM_011509168.1 XP_011507470.1 17 2244
XM_011509169.1 XP_011507471.1 16 2033
XM_011509170.1 XP_011507472.1 17 2240
XM_011509171.1 XP_011507473.1 16 2058
XM_011509174.1 XP_011507476.1 17 2240
XM_011509175.1 XP_011507477.1 17 2240
XM_011509176.1 XP_011507478.1 17 2240
XM_011509178.2 XP_011507480.1 17 2240
XM_011509179.2 XP_011507481.1 12 1348
XM_011509180.1 XP_011507482.1 17 2240
XM_011509181.2 XP_011507483.1 7 857
XM_017000257.1 XP_016855746.1 11 1208
XM_017000258.1 XP_016855747.1 11 1240
XM_017000259.1 XP_016855748.1 3 410
GenBank mRNA AK057502.1 - 16 1771
AK302972.1 - 18 2383
AL833914.1 - 15 1688
BC146823.1 - 16 1901 8860438
BX647935.1 - 17 2262

View Full Product Details