Gene Transcripts

Gene Symbol
EMID1
Entrez Gene ID
129080
Gene Name
EMI domain containing 1
Gene Aliases
EMI5, EMU1
Chromosome Location
Chr. 22: 29205912 - 29259597 on Build GRCh38
UniGene ID
Hs.289106
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267895.1 NP_001254824.1 8, 9 955
NM_133455.3 NP_597712.2 8, 9 961
XM_005261328.2 XP_005261385.1 8, 9 956
XM_005261329.2 XP_005261386.1 7, 8 866
XM_011529868.2 XP_011528170.1 9, 10 967
XM_011529869.2 XP_011528171.1 9, 10 966
XM_011529870.2 XP_011528172.1 9, 10 966
XM_011529871.2 XP_011528173.1 8, 9 885
XM_011529872.2 XP_011528174.1 9, 10 967
XM_011529873.2 XP_011528175.1 9, 10 967
XM_011529874.2 XP_011528176.1 10, 11 3602
XM_011529875.2 XP_011528177.1 5, 6 506
XM_011529876.1 XP_011528178.1 7, 8 599
XM_017028589.1 XP_016884078.1 9, 10 4196
GenBank mRNA AJ416090.1 - 8, 9 828
AL831910.1 - 1 1708
BC046358.1 - 8, 9 909 5770546
CR456407.1 - 8, 9 844

View Full Product Details