Gene Transcripts

Gene Symbol
PLEKHH2
Entrez Gene ID
130271
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H2
Gene Aliases
PLEKHH1L
Chromosome Location
Chr. 2: 43637300 - 43767987 on Build GRCh38
UniGene ID
Hs.164162
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172069.3 NP_742066.2 17 2748
XM_017003351.1 XP_016858840.1 16 2735
XM_017003352.1 XP_016858841.1 17 2748
XM_017003353.1 XP_016858842.1 18 2859
GenBank mRNA AB095948.1 - 15 2534
AK126308.1 - 17 2744
AL832207.1 - 17 2741
AL833400.1 - 10 1051
AL833546.1 - 17 4594

View Full Product Details